Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc77 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_045558.1 | 5 - 6 | 765 | 93 | ||
GenBank mRNA | AK031590.1 | - | 5 - 6 | 766 | 93 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | beta-1,4-N-acetyl-galactosaminyl transferase 3 | B4galnt3 |
AB114826, B4GalNAcT3, C330047A21 |
-43.1kb | Sense | Chr. 6 - 120200753 - 120295005 on NCBI Build GRCm38 |
Downstream | lysine (K)-specific demethylase 5A | Kdm5a |
AA409370, C76986, Jarid1a, RBP2, Rbbp2 |
22.0kb | Anti-sense | Chr. 6 - 120364099 - 120444574 on NCBI Build GRCm38 |
Set Membership: |
Amplicon length between 86 and 100 Mus musculus Probe spans exons |