Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Crem | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001110850.2 | NP_001104320.1 | 4 - 5 | 456 | 82 | |
NM_001110858.2 | NP_001104328.1 | 4 - 5 | 456 | 82 | ||
NM_001271503.1 | NP_001258432.1 | 5 - 6 | 603 | 82 | ||
NM_001271504.1 | NP_001258433.1 | 5 - 6 | 603 | 82 | ||
NR_073193.1 | 6 - 7 | 699 | 82 | |||
GenBank mRNA | AY738715.1 | - | 4 - 5 | 519 | 82 | |
AY738716.1 | - | 3 - 4 | 372 | 82 | ||
AY738717.1 | - | 4 - 5 | 519 | 82 | ||
AY738720.1 | - | 5 - 6 | 615 | 82 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mus musculus Probe spans exons |