Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Btbd8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001255991.1 | NP_001242920.1 | 7 - 8 | 924 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Mus musculus Probe spans exons |