Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1100012823.mm.1 |
NM_016686.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100012823.mm.1 |
TC1100001361.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100012822.mm.1 |
NM_016686.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100012822.mm.1 |
TC1100001361.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100012819.mm.1 |
NM_016686.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100012819.mm.1 |
TC1100001361.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100012821.mm.1 |
NM_016686.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100012821.mm.1 |
TC1100001361.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100012820.mm.1 |
NM_016686.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100012820.mm.1 |
TC1100001361.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100012824.mm.1 |
NM_016686.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100012824.mm.1 |
TC1100001361.mm.1 |
MTA TC |