Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vezf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_016686.4 | NP_057895.2 | 5 - 6 | 1253 | 79 | |
GenBank mRNA | AF104410.3 | - | 5 - 6 | 1253 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 3' Most Mus musculus Probe spans exons |