Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001301737.1 | NP_001288666.1 | 16 - 17 | 1904 | 105 | |
NM_010065.3 | NP_034195.2 | 15 - 16 | 1892 | 105 | ||
NR_125959.1 | 16 - 17 | 1904 | 105 | |||
GenBank mRNA | AK144142.1 | - | 15 - 16 | 1843 | 105 | |
AK220483.1 | - | 16 - 17 | 1846 | 105 | ||
BC034679.1 | - | 16 - 17 | 1858 | 4506865 | 105 | |
BC058623.1 | - | 16 - 17 | 1838 | 5706803 | 105 | |
BM950315.1 | - | 2 - 3 | 297 | 105 | ||
CO425806.1 | - | 2 - 3 | 223 | 105 | ||
L31395.1 | - | 16 - 17 | 1801 | 105 | ||
L31396.1 | - | 11 - 12 | 1073 | 105 | ||
L31397.1 | - | 15 - 16 | 1845 | 105 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |