Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hnrnpk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001301340.1 | NP_001288269.1 | 16 | 2633 | 104 | |
NM_001301341.1 | NP_001288270.1 | 18 | 2760 | 104 | ||
NM_001301343.1 | NP_001288272.1 | 16 | 2619 | 104 | ||
NM_001301344.1 | NP_001288273.1 | 15 | 2561 | 104 | ||
NM_001301345.1 | NP_001288274.1 | 15 | 2501 | 104 | ||
NM_025279.3 | NP_079555.1 | 16 | 2573 | 104 | ||
GenBank mRNA | AK128891.1 | - | 16 | 2570 | 104 | |
AK135519.1 | - | 16 | 2546 | 104 | ||
AK151689.1 | - | 15 | 2514 | 104 | ||
AK152927.1 | - | 15 | 2515 | 104 | ||
AK160887.1 | - | 16 | 2574 | 104 | ||
AK166656.1 | - | 16 | 2637 | 104 | ||
AK166927.1 | - | 16 | 2591 | 104 | ||
AK167600.1 | - | 16 | 2563 | 104 | ||
AK168918.1 | - | 12 | 2474 | 105 | ||
AK169064.1 | - | 16 | 2622 | 104 | ||
BC089328.1 | - | 16 | 2557 | 6402614 | 104 | |
BE993983.1 | - | 1 | 104 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Apoptosis 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus Both primers and probe map within a single exon |