Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdlim7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_104287.1 | 6 - 7 | 502 | 81 | ||
GenBank mRNA | AK003019.1 | - | 6 - 7 | 459 | 81 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | microRNA 6945 | Mir6945 |
Gm27779, mmu-mir-6945 |
Sense | Chr. 13 - 55507625 - 55507692 on NCBI Build GRCm38 | |
Downstream | docking protein 3 | Dok3 |
AI450713, Dokl |
15.4kb | Sense | Chr. 13 - 55523235 - 55528538 on NCBI Build GRCm38 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Mus musculus Probe spans exons |