Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnf41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_104288.1 | 4 - 5 | 756 | 103 | ||
GenBank mRNA | AK045760.1 | - | 4 - 5 | 756 | 103 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | nucleic acid binding protein 2 | Nabp2 |
2610036N15Rik, Obfc2b |
-23.8kb | Anti-sense | Chr. 10 - 128401394 - 128411487 on NCBI Build GRCm38 |
Downstream | microRNA 8105 | Mir8105 |
Gm27346, mmu-mir-8105 |
22.1kb | Sense | Chr. 10 - 128458680 - 128458768 on NCBI Build GRCm38 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mus musculus Probe spans exons |