Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cklf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_104433.1 | 4 - 5 | 605 | 76 | ||
GenBank mRNA | AF401531.2 | - | 4 - 5 | 510 | 76 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
thymidine kinase 2, mitochondrial |
Tk2 |
AU024611, mt-TK |
-13.6kb | Anti-sense | Chr. 8 - 104226691 - 104248574 on NCBI Build GRCm38 |
Downstream | CKLF-like MARVEL transmembrane domain containing 2A | Cmtm2a |
1700001K04Rik, 1700041N15Rik, 1700063K20Rik, AI449770, ARR19, C32, CKLF3, CKLF4, CKLF5, Cklf, Cklf1, Cklfsf2-1b, Cklfsf2a, UCK-1 |
17.6kb | Anti-sense | Chr. 8 - 104281042 - 104293181 on NCBI Build GRCm38 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mus musculus Probe spans exons |