Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1000006219.mm.1 |
NR_110342.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000006219.mm.1 |
TC1000000837.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000006220.mm.1 |
NR_110342.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000006220.mm.1 |
TC1000000837.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000006236.mm.1 |
NM_001037741.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000006236.mm.1 |
NM_008162.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000006236.mm.1 |
NR_110342.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000006236.mm.1 |
TC1000000837.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000006240.mm.1 |
NM_001037741.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000006240.mm.1 |
NM_008162.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000006240.mm.1 |
NR_110342.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000006240.mm.1 |
TC1000000837.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000006238.mm.1 |
NM_001037741.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000006238.mm.1 |
NM_008162.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000006238.mm.1 |
NR_110342.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000006238.mm.1 |
TC1000000837.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000006241.mm.1 |
NM_001037741.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000006241.mm.1 |
NM_008162.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000006241.mm.1 |
NR_110342.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000006241.mm.1 |
TC1000000837.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000006234.mm.1 |
NM_001037741.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000006234.mm.1 |
NM_008162.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000006234.mm.1 |
NR_110342.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000006234.mm.1 |
TC1000000837.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000006231.mm.1 |
NM_001037741.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000006231.mm.1 |
NM_008162.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000006231.mm.1 |
NR_110342.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000006231.mm.1 |
TC1000000837.mm.1 |
MTA TC |