Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sh3yl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_110343.1 | 1 - 2 | 557 | 114 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
acid phosphatase 1, soluble |
Acp1 |
4632432E04Rik, AI427468, Acp-1, LMW-PTP |
-48.0 | Anti-sense | Chr. 12 - 30893326 - 30911621 on NCBI Build GRCm38 |
Downstream |
family with sequence similarity 110, member C |
Fam110c |
9030611O19Rik, AU019489 |
160.9kb | Sense | Chr. 12 - 31073968 - 31079940 on NCBI Build GRCm38 |
Set Membership: |
Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |