Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ranbp17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_110973.1 | 15 - 16 | 1954 | 107 | ||
GenBank mRNA | BC033503.1 | - | 15 - 16 | 1848 | 107 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
T cell leukemia, homeobox 3 |
Tlx3 |
Hox11l2, Rnx, Tlx1l2 |
-125.9kb | Sense | Chr. 11 - 33200752 - 33203588 on NCBI Build GRCm38 |
Downstream |
gamma-aminobutyric acid (GABA) A receptor, pi |
Gabrp | 146.6kb | Sense | Chr. 11 - 33550781 - 33578978 on NCBI Build GRCm38 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |