Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010554.4 | NP_034684.2 | 4 - 5 | 393 | 70 | |
GenBank mRNA | AK156449.1 | - | 4 - 5 | 394 | 70 | |
BC003727.1 | - | 3 - 4 | 362 | 3599550 | 70 | |
X01450.1 | - | 4 - 5 | 393 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Inventoried Probe spans exons |