Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZFPM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027973106.1 | XP_027828907.1 | 6 - 7 | 760 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Ovis aries |