Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TADA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015102307.2 | XP_014957793.1 | 4 - 5 | 634 | 86 | |
XM_027958097.1 | XP_027813898.1 | 4 - 5 | 634 | 86 | ||
XM_027958098.1 | XP_027813899.1 | 4 - 5 | 631 | 86 | ||
XM_027958099.1 | XP_027813900.1 | 4 - 5 | 631 | 86 | ||
XM_027958100.1 | XP_027813901.1 | 4 - 5 | 837 | 86 | ||
XM_027958101.1 | XP_027813902.1 | 4 - 5 | 631 | 86 | ||
XM_027958102.1 | XP_027813903.1 | 4 - 5 | 637 | 86 | ||
XM_027958103.1 | XP_027813904.1 | 4 - 5 | 635 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Ovis aries |