Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GABARAPL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012175401.3 | XP_012030791.1 | 3 - 4 | 571 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Ovis aries |