Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATP1B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009796.1 | NP_001009796.1 | 3 - 4 | 753 | 98 | |
XM_027975567.1 | XP_027831368.1 | 2 - 3 | 710 | 98 | ||
GenBank mRNA | X03883.1 | - | 753 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Ovis aries |