Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LUM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004006251.3 | XP_004006300.1 | 2 - 3 | 1235 | 98 | |
XM_012174076.3 | XP_012029466.1 | 2 - 3 | 990 | 98 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Ovis aries |