Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TACR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015104406.2 | XP_014959892.2 | 1 - 2 | 872 | 99 |
Set Membership: |
Amplicon length between 86 and 100 G-protein coupled receptor Ovis aries |