Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRKACA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009234.1 | NP_001009234.1 | 4 - 5 | 348 | 77 | |
XM_027969189.1 | XP_027824990.1 | 4 - 5 | 338 | 77 | ||
GenBank mRNA | AF238979.1 | - | 4 - 5 | 348 | 77 | |
AF238980.1 | - | 4 - 5 | 343 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Ovis aries |