Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC101110178 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027980021.1 | XP_027835822.1 | 6 - 7 | 738 | 84 | |
XM_027980022.1 | XP_027835823.1 | 6 - 7 | 738 | 84 | ||
XM_027980023.1 | XP_027835824.1 | 6 - 7 | 774 | 84 | ||
XM_027980024.1 | XP_027835825.1 | 6 - 7 | 738 | 84 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Ovis aries |