Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AQP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009279.1 | NP_001009279.1 | 4 - 5 | 707 | 72 | |
XM_012103242.3 | XP_011958632.1 | 4 - 5 | 905 | 72 | ||
XM_012103243.3 | XP_011958633.1 | 4 - 5 | 964 | 72 | ||
XM_012103244.3 | XP_011958634.1 | 4 - 5 | 875 | 72 | ||
GenBank mRNA | AY177612.1 | - | 4 - 5 | 698 | 72 | |
AY206465.1 | - | 3 - 4 | 626 | 72 | ||
AY304480.1 | - | 4 - 5 | 707 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Ovis aries |