Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLDN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001185016.1 | NP_001171945.1 | 3 - 4 | 468 | 97 | |
GenBank mRNA | DQ152937.1 | - | 1 - 2 | 91 | 97 | |
HM117762.1 | - | 3 - 4 | 468 | 97 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Ovis aries |