Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSMC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027972123.1 | XP_027827924.1 | 5 - 6 | 666 | 51 | |
XM_027972124.1 | XP_027827925.1 | 5 - 6 | 525 | 51 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Ovis aries |