Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WNT16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004008012.4 | XP_004008061.2 | 2 - 3 | 752 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Ovis aries |