Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BSCL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012102042.2 | XP_011957432.1 | 3 - 4 | 616 | 59 | |
XM_012102044.3 | XP_011957434.1 | 3 - 4 | 631 | 59 | ||
XM_012102045.3 | XP_011957435.1 | 3 - 4 | 650 | 59 | ||
XM_012102047.2 | XP_011957437.1 | 3 - 4 | 985 | 59 | ||
XM_012102048.3 | XP_011957438.1 | 2 - 3 | 384 | 59 | ||
XM_012102049.3 | XP_011957439.1 | 2 - 3 | 476 | 59 | ||
XM_027960097.1 | XP_027815898.1 | 3 - 4 | 614 | 59 | ||
XM_027960098.1 | XP_027815899.1 | 3 - 4 | 614 | 59 | ||
XM_027960099.1 | XP_027815900.1 | 3 - 4 | 614 | 59 |