Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RTN4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001145178.2 | NP_001138650.2 | 4 - 5 | 695 | 83 | |
NM_001375595.1 | NP_001362524.1 | 6 - 7 | 3651 | 83 | ||
XM_015093765.2 | XP_014949251.1 | 4 - 5 | 501 | 83 | ||
XM_027965718.1 | XP_027821519.1 | 4 - 5 | 1170 | 83 | ||
GenBank mRNA | FJ546074.1 | - | 4 - 5 | 594 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Ovis aries |