Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRDX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004001918.3 | XP_004001967.1 | 3 - 4 | 358 | 68 | |
XM_012126652.3 | XP_011982042.2 | 3 - 4 | 338 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Ovis aries |