Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSD11B2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009460.1 | NP_001009460.1 | 1 - 2 | 362 | 50 | |
GenBank mRNA | U14128.1 | - | 1 - 2 | 362 | 50 |
Set Membership: |
Amplicon length less than or equal to 70 Ovis aries |