Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001256817.1 | NP_001243746.1 | 1 - 2 | 180 | 118 | |
GenBank mRNA | HE652877.1 | - | 1 - 2 | 180 | 118 |
Set Membership: |
Amplicon length greater than or equal to 101 Ovis aries |