Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OS9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004006523.2 | XP_004006572.1 | 1 - 2 | 196 | 73 | |
XM_004006525.2 | XP_004006574.1 | 1 - 2 | 197 | 73 | ||
XM_012174649.2 | XP_012030039.1 | 1 - 2 | 197 | 73 | ||
XM_012174650.2 | XP_012030040.1 | 1 - 2 | 197 | 73 | ||
XM_012174651.2 | XP_012030041.1 | 1 - 2 | 197 | 73 | ||
XM_012174652.2 | XP_012030042.1 | 1 - 2 | 197 | 73 | ||
XM_027967513.1 | XP_027823314.1 | 1 - 2 | 197 | 73 |