Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GATD3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001162560.1 | NP_001156032.1 | 3 - 4 | 325 | 78 | |
GenBank mRNA | FJ969404.1 | - | 3 - 4 | 325 | 78 |