Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNAPC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027969735.1 | XP_027825536.1 | 4 - 5 | 1291 | 83 | |
XM_027969736.1 | XP_027825537.1 | 4 - 5 | 1419 | 83 |