Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SERPINC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009393.1 | NP_001009393.1 | 3 - 4 | 680 | 118 | |
GenBank mRNA | X68287.1 | - | 680 | 118 |
Set Membership: |
Amplicon length greater than or equal to 101 Ovis aries |