Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TCTA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004018461.4 | XP_004018510.1 | 1 - 2 | 446 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Ovis aries |