Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SUPT4H1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004012398.4 | XP_004012447.1 | 4 - 5 | 637 | 80 | |
XM_027975360.1 | XP_027831161.1 | 3 - 4 | 315 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Ovis aries |