Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MLX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015098794.2 | XP_014954280.2 | 2 - 3 | 263 | 59 | |
XM_027974498.1 | XP_027830299.1 | 2 - 3 | 138 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Ovis aries |