Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: XPA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004004232.4 | XP_004004281.1 | 1 - 2 | 197 | 92 | |
XM_012120695.3 | XP_011976085.1 | 1 - 2 | 187 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Ovis aries |