Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TIMM9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012181724.3 | XP_012037114.1 | 3 - 4 | 291 | 95 | |
XM_012181726.3 | XP_012037116.1 | 4 - 5 | 370 | 95 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Ovis aries |