Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC101120043 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015104035.2 | XP_014959521.2 | 3 - 4 | 655 | 93 | |
XM_027961464.1 | XP_027817265.1 | 3 - 4 | 686 | 93 | ||
XM_027961465.1 | XP_027817266.1 | 3 - 4 | 615 | 93 | ||
XM_027961466.1 | XP_027817267.1 | 3 - 4 | 649 | 93 | ||
XM_027961468.1 | XP_027817269.1 | 2 - 3 | 550 | 93 | ||
XM_027961469.1 | XP_027817270.1 | 2 - 3 | 594 | 93 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Ovis aries |