Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001161891.1 | NP_001155363.1 | 2 - 3 | 272 | 94 | |
GenBank mRNA | FJ969421.1 | - | 2 - 3 | 272 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Ovis aries |