Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC25A33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127271.1 | NP_001120743.1 | 5 - 6 | 488 | 58 | |
GenBank mRNA | EU626216.1 | - | 5 - 6 | 488 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Ovis aries |