Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HCST | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012190204.3 | XP_012045594.1 | 3 - 4 | 186 | 99 | |
XM_012190205.3 | XP_012045595.1 | 3 - 4 | 223 | 99 | ||
XM_012190206.3 | XP_012045596.1 | 2 - 3 | 1022 | 99 |