Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CMC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012107036.2 | XP_011962426.1 | 2 - 3 | 352 | 81 | |
XM_027962990.1 | XP_027818791.1 | 3 - 4 | 157 | 81 | ||
XM_027962991.1 | XP_027818792.1 | 2 - 3 | 159 | 81 | ||
XM_027962992.1 | XP_027818793.1 | 2 - 3 | 163 | 81 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Ovis aries |