Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AURKB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004012681.4 | XP_004012730.2 | 3 - 4 | 261 | 63 | |
XM_012185693.3 | XP_012041083.2 | 3 - 4 | 279 | 63 | ||
XM_027974924.1 | XP_027830725.1 | 2 - 3 | 199 | 63 | ||
XM_027974925.1 | XP_027830726.1 | 3 - 4 | 279 | 63 | ||
XM_027974926.1 | XP_027830727.1 | 3 - 4 | 236 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Ovis aries |