Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027967251.1 | XP_027823052.1 | 3 - 4 | 1800 | 60 | |
XM_027967253.1 | XP_027823054.1 | 3 - 4 | 2117 | 60 | ||
XM_027967254.1 | XP_027823055.1 | 3 - 4 | 1656 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Ovis aries |