Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LLGL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015098677.2 | XP_014954163.2 | 1 - 2 | 125 | 104 | |
XM_027974801.1 | XP_027830602.1 | 1 - 2 | 116 | 104 | ||
XM_027974802.1 | XP_027830603.1 | 1 - 2 | 116 | 104 | ||
XM_027974803.1 | XP_027830604.1 | 1 - 2 | 116 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Ovis aries |