Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SAE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004015334.4 | XP_004015383.1 | 3 - 4 | 498 | 90 | |
XM_012190581.3 | XP_012045971.1 | 3 - 4 | 451 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Ovis aries |