Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004006941.4 | XP_004006990.1 | 2 - 3 | 735 | 66 | |
XM_012175528.3 | XP_012030918.1 | 2 - 3 | 735 | 66 |